Orders LinkContactUsLinkComponent
Slide Boxes
Slide Boxes
Transparent, PC, 50, 172×83×31 mm
Catalog # GLWAK51
Supplier:   GLW
Slide Boxes
Catalog # GLWAK51
Supplier:  GLW
Slide Boxes
Slide Boxes
Transparent, PC, 50, 172×83×31 mm
Price
10,26 €
Log in for availability
1 * 1 ST
Product Details & Documents
High quality slide storage boxes for microscopy slides.
Used for storing, transporting and freezing in science, industry and biotechnology.
Specifications
  • No. of slides:
    50
  • Colour:
    Transparent
  • Material:
    PC
  • Seq:
    10
  • W×D×H:
    172×83×31 mm
Accessories
Product Information
  • Catalog #
    Colour
    Material
    Availability
    Price
  • Divider for slides and coverslips of special sizes
    Divider for slides and coverslips of special sizes
    GLWAZK20
    Grey
    PS
    Log in for availability
    1 * 1 ST - 2,80 €
Product Information
  • Catalog #
    Colour
    Material
    Availability
    Price
  • Divider for slides and coverslips of special sizes
    Divider for slides and coverslips of special sizes
    GLWAZK15
    White
    PS
    Log in for availability
    1 * 1 Un. - 5,94 €