Order Entry
Belgium
Orders LinkContactUsLinkComponent
pH buffer, single-use, SINGLET™

pH buffer, single-use, SINGLET™

pH buffer solution, single-use, 10,01, ±0,02, 25 °C, Foil pouch, 20×20 ml pouches, 20
Catalog # HACH2770220
Supplier:   Hach
pH buffer, single-use, SINGLET™
Catalog # HACH2770220
Supplier:  Hach
CAS Number:  
pH buffer, single-use, SINGLET™
pH buffer, single-use, SINGLET™

pH buffer solution, single-use, 10,01, ±0,02, 25 °C, Foil pouch, 20×20 ml pouches, 20

CAS Number:  
Price
50,70 €
Log in for availability
1 * 20 ST
Product Details & Documents

These buffers solutions packs are used for pH meter calibration. The sealed foil pouches can be transported easily and safely without any spillage and also prevents contamination or exposure to air of the solutions.

  • Ideal for on-site analyses
  • Easy to use
  • Accuracy pH: ±0,02 at 25 °C
  • Traceable to NIST

Documents

Specifications

Specifications

  • Pack type:
    Foil pouch
  • Pk:
    20
  • Reference temperature:
    25 °C
  • Description:
    pH buffer solution, single-use
  • pH accuracy:
    ±0,02
  • Pk Info:
    20×20 ml pouches
  • pH value:
    10,01

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.