Orders LinkContactUsLinkComponent
Test kits, digital titration method
Test kits, digital titration method
Chloride CDS-DT: Silver nitrate method, 10 - 10000 mg/l, 1 KIT
N° de catalogue HACH2580600
Fournisseur:   Hach
Test kits, digital titration method
N° de catalogue HACH2580600
Fournisseur:  Hach
Numéro CAS:  
Test kits, digital titration method
Test kits, digital titration method
Chloride CDS-DT: Silver nitrate method, 10 - 10000 mg/l, 1 KIT
Numéro CAS:  
Prix
642,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
Digital titration provides a more precise and accurate method than the drop test method.
  • Reagents are pre-prepared
  • Exact comparison of colour
  • Rugged carrying case for analysis in the field
Caution: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Specifications
Specifications
  • Pk:
    1 KIT
  • Description:
    Chloride CDS-DT: Silver nitrate method
  • Range:
    10 - 10000 mg/l