Order Entry
Belgium
Orders LinkContactUsLinkComponent
Iron standard solution

Iron standard solution

Iron standard solution, 500 ml (NIST), 1 mg/l as Fe, 500 mL
Catalog # HACH13949
Supplier:   Hach
Iron standard solution
Catalog # HACH13949
Supplier:  Hach
CAS Number:  
Iron standard solution
Iron standard solution

Iron standard solution, 500 ml (NIST), 1 mg/l as Fe, 500 mL

CAS Number:  
Price
46,20 €
Log in for availability
1 * 500 mL
Product Details & Documents

For accuracy checks in iron determinations.

  • Traceable to NIST
Specifications

Specifications

  • Concentration:
    1 mg/l as Fe
  • Pk:
    500 mL
  • Description:
    Iron standard solution, 500 ml (NIST)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.