Orders LinkContactUsLinkComponent
Accessories for parallel synthesis, Syncore® Polyvap, Syncore® Analyst
Glass sample tubes for rack R-6 polyvap, round bottom (Ø 75 mm), height 175 mm, for working volume up to 250 ml
Catalog # BUCH038486
Supplier:   BUCHI
Accessories for parallel synthesis, Syncore® Polyvap, Syncore® Analyst
Catalog # BUCH038486
Supplier:  BUCHI
Accessories for parallel synthesis, Syncore® Polyvap, Syncore® Analyst
Glass sample tubes for rack R-6 polyvap, round bottom (Ø 75 mm), height 175 mm, for working volume up to 250 ml
Price
530,00 €
Log in for availability
1 * 6 ST
Product Details & Documents
Specifications
  • Description:
    Glass sample tubes for rack R-6 polyvap, round bottom (Ø 75 mm), height 175 mm, for working volume up to 250 ml
  • Seq:
    02