Order Entry
Belgium
Orders LinkContactUsLinkComponent
Staining dishes, WHEATON®

Staining dishes, WHEATON®

Staining dish, only, 95×76×64 mm
Catalog # 631-2501
Supplier:   DWK Life Sciences
Staining dishes, WHEATON®
Catalog # 631-2501
Supplier:  DWK Life Sciences
Staining dishes, WHEATON®
Staining dishes, WHEATON®

Staining dish, only, 95×76×64 mm

Price
175,00 €
Log in for availability
1 * 3 ST
Product Details & Documents

Soda-lime glass and stainless steel.

  • Removable glass slide rack has an open bottom to facilitate rapid immersion and drainage, reducing carry-over

Standard for manual staining procedures. The rack holds 10 single slides, 19 slides arranged alternately straight across and diagonally, or 20 slides back to back of standard size 75×25 mm, 75×38 mm and 75×51 mm.

Documents

Specifications
  • Description:
    Staining dish, only
  • W×D×H:
    95×76×64 mm