Order Entry
Belgium
Orders LinkContactUsLinkComponent
Tissue processing/embedding cassettes, Histosette® I, M490

Tissue processing/embedding cassettes, Histosette® I, M490

Purple
Catalog # 720-0884
Supplier:   Simport Scientific
Tissue processing/embedding cassettes, Histosette® I, M490
Catalog # 720-0884
Supplier:  Simport Scientific
Tissue processing/embedding cassettes, Histosette® I, M490
Tissue processing/embedding cassettes, Histosette® I, M490

Purple

Price
163,00 €
Log in for availability
1 * 1.500 ST
Product Details & Documents

Acetal polymer.

  • Resistant to histological solvents
  • Flow-through slots maximise fluid exchange and ensure proper drainage
  • Integral lid eliminates the need for separate steel lids
  • 30° writing angle
Specifications
  • Colour:
    Purple