Order Entry
Belgium
Orders LinkContactUsLinkComponent
Cuvettes for Amelung®

Cuvettes for Amelung®

Amelung® cuvettes, 1 ml, 18,65/11,25×22,8 mm
Catalog # KART82631
Supplier:   KARTELL
Cuvettes for Amelung®
Catalog # KART82631
Supplier:  KARTELL
Cuvettes for Amelung®
Cuvettes for Amelung®

Amelung® cuvettes, 1 ml, 18,65/11,25×22,8 mm

Price
51,50 €
Log in for availability
1 * 1.000 ST
Product Details & Documents

Polystyrene cuvettes designed for the determination of coagulation time.

Specifications
  • Ø×H:
    18,65/11,25×22,8 mm
  • Volume:
    1 ml
  • Description:
    Amelung® cuvettes
  • Seq:
    001