Forceps
Blunt, 253 mm
Catalog # 232-2003
Supplier: KARTELL
Jump to:
Product Details & Documents
Polyoxymethylene (POM), yellow. Multipurpose forceps for a wide range of applications.
- Good chemical resistance, not suitable for use with strong acids and solvents
- Autoclavable
- Robust, pliable, self-sprung
Specifications
- Type:Blunt
- Length:253 mm