Orders LinkContactUsLinkComponent
Forceps
Forceps
Blunt, 118 mm
Catalog # 232-2000
Supplier:   KARTELL
Forceps
Catalog # 232-2000
Supplier:  KARTELL
Forceps
Forceps
Blunt, 118 mm
Price
32,70 €
Log in for availability
1 * 5 ST
Product Details & Documents
Polyoxymethylene (POM), yellow. Multipurpose forceps for a wide range of applications.
  • Good chemical resistance, not suitable for use with strong acids and solvents
  • Autoclavable
  • Robust, pliable, self-sprung
Specifications
  • Type:
    Blunt
  • Length:
    118 mm