Order Entry
Belgium
Orders LinkContactUsLinkComponent
Flat spatula, conical

Flat spatula, conical

Stainless steel, 120 mm, 15/25 mm
Catalog # NICK2122
Supplier:   NICKEL ELECTRO
Flat spatula, conical
Catalog # NICK2122
Supplier:  NICKEL ELECTRO
Flat spatula, conical
Flat spatula, conical

Stainless steel, 120 mm, 15/25 mm

Price
7,30 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel or pure nickel (99,6%). Multipurpose spatula for a wide range of uses.

  • Excellent chemical resistance
  • Excellent thermal resistance
  • Smooth surfaces for fast, easy cleaning
Specifications
  • Description:
    Stainless steel
  • Length:
    120 mm
  • Blade width:
    15/25 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.