Orders LinkContactUsLinkComponent
Dissecting trays
Dissecting trays
500×350×50 mm
Catalog # NICK698
Supplier:   NICKEL ELECTRO
Dissecting trays
Catalog # NICK698
Supplier:  NICKEL ELECTRO
Dissecting trays
Dissecting trays
500×350×50 mm
Price
145,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel.
  • Very good chemical resistance
  • Very good temperature resistance
  • Easy handling
For use in laboratories and schools.
Specifications
  • W×D×H:
    500×350×50 mm