Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Storage Boxes

Red, 595×395×320 mm
  216-2877
Storage Boxes
  216-2877
Storage Boxes
Storage Boxes

Red, 595×395×320 mm

 
 
   
1 * 1 ST
 

PP, coloured.
Ideal for a wide range of uses. With tightly fitting lid.

  • Very good chemical resistance
  • Very good thermal resistance
  • Very sturdy and economical
 
  • Colour:
    Red
  • Seq:
    14
  • W×D×H:
    595×395×320 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.