Order Entry
Belgium
Orders LinkContactUsLinkComponent
Storage Boxes

Storage Boxes

Blue, 395×295×160 mm
Catalog # 216-2868
Supplier:   Gilac PROFESSIONNEL
Storage Boxes
Catalog # 216-2868
Storage Boxes
Storage Boxes

Blue, 395×295×160 mm

Price
56,90 €
Log in for availability
1 * 1 ST
Product Details & Documents

PP, coloured.
Ideal for a wide range of uses. With tightly fitting lid.

  • Very good chemical resistance
  • Very good thermal resistance
  • Very sturdy and economical
Specifications
  • Colour:
    Blue
  • Seq:
    02
  • W×D×H:
    395×295×160 mm