Cooling systems, CoolSafe™
CoolSafe™ systems, including CoolSafe box with lid, cooling chamber and each one cool brick 1 °C and -10 °C, 0,2/0,5 and 1,5 ml tubes, 79 (20 for 1,5 ml/35 for 0,5 ml/24 for 0,2 ml), 140×216×165 mm
DIVECSAF-5000
Polystyrene cool safe box, aluminium cooling chamber.
- Durable
- Temperature stable
- Corrosion resistant
Cooling system designed for 0,2 ml tubes (strips), 96 and 384 well microplates, 0,5, 1,5 and 5,0 ml tubes. Cool bricks with different temperature zones 1 or −10 °C.
- Pour:0,2/0,5 and 1,5 ml tubes
- Nb de places:79 (20 for 1,5 ml/35 for 0,5 ml/24 for 0,2 ml)
- Désignation:CoolSafe™ systems, including CoolSafe box with lid, cooling chamber and each one cool brick 1 °C and -10 °C
- Dimensions:140×216×165 mm
- Seq:19



