Orders LinkContactUsLinkComponent
Accessories for on-line multi-parameter systems, IQ sensor net
Accessories for on-line multi-parameter systems, IQ sensor net
Tetracon 700 IQ Digital, 4-pole conductivity cell for waste water, with integrated temperature sensor, without SACIQ cable
Catalog # 708-0045
Supplier:   WTW ONLINE
Accessories for on-line multi-parameter systems, IQ sensor net
Catalog # 708-0045
Supplier:  WTW ONLINE
Accessories for on-line multi-parameter systems, IQ sensor net
Accessories for on-line multi-parameter systems, IQ sensor net
Tetracon 700 IQ Digital, 4-pole conductivity cell for waste water, with integrated temperature sensor, without SACIQ cable
Price
1 590,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Tetracon 700 IQ Digital, 4-pole conductivity cell for waste water, with integrated temperature sensor, without SACIQ cable
  • Seq:
    10