Orders LinkContactUsLinkComponent
Filter sets
Filter sets
Filter set 31, excitation BP 565/30, beam splitter FT 585, emission BP 620/60
Catalog # ZEIS0000001031350
Supplier:   ZEISS
Filter sets
Catalog # ZEIS0000001031350
Supplier:  ZEISS
Filter sets
Filter set 31, excitation BP 565/30, beam splitter FT 585, emission BP 620/60
Price
1 754,87 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Filter set 31, excitation BP 565/30, beam splitter FT 585, emission BP 620/60