Objectives
Epi-Planachromatic, N.A. of 0,13, WD of 17,1 mm
Catalog # ZEIS4429240000000
Supplier: ZEISS
Jump to:
Product Details & Documents
Specifications
- Magnification:5×
- Description:Epi-Planachromatic, N.A. of 0,13, WD of 17,1 mm