Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Multi-way stopcocks, Discofix®

Three-way stopcocks with 250 mm extension
  BRAU4098218
 :   B. Braun
Multi-way stopcocks, Discofix®
  BRAU4098218
 :  B. Braun
Multi-way stopcocks, Discofix®
Multi-way stopcocks, Discofix®

Three-way stopcocks with 250 mm extension

 
239,00 €
   
1 * 50 ST
 

Suitable for all pressure infusion systems <2 bar.

  • Continuous flow channels for accurate pressure monitoring
  • Swivel lock can be moved axially or radially for fast and secure connection
  • PVC and latex-free

Suitable for all pressure infusion systems <2 bar.

 
  • Description:
    Three-way stopcocks with 250 mm extension