Order Entry
Belgium
Orders LinkContactUsLinkComponent
Measuring scoops

Measuring scoops

100 ml, 200 mm
  231-0068
 :   KARTELL
Measuring scoops
  231-0068
 :  KARTELL
Measuring scoops
Measuring scoops

100 ml, 200 mm

 
77,50 €
   
1 * 12 ST
 

PP, white. With flat, bevelled end and reinforced handle.

  • Excellent chemical resistance, unbreakable
  • Heat resistant up to 130 °C, autoclavable
  • The capacity of the scoop is stamped into the handle for easy reading
 
  • Capacity:
    100 ml
  • Length:
    200 mm
  • Seq:
    08
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.