FILTER 0.2µ AUTOCLAVABLE SMART DILUTOR 1 * 2 items
Catalog # IULA8182
Supplier: IUL Instruments SA
Frequently Bought Together
VWR® Counter-Pen™ Cell Counter
Supplier: VWR Collection
ThermoMixer® C Thermomixer
Supplier: EPPENDORF
Accessories for OxiTop® and OxiTop® Control
Supplier: WTW
Accessories for Mettler balances
Supplier: Mettler - Toledo
SmartBlocks for ThermoMixer® C and ThermoStat C
Supplier: EPPENDORF
Electrode stand with swivel arm, Eutech
Supplier: EUTECH
Flow indicator paddle wheel
Supplier: Brand
Titration beakers
Supplier: Mettler - Toledo
pH/mV/°C meters, handheld, pH 3110 / 3310
Supplier: WTW
Accessories for flow cups with interchangeable nozzle
Supplier: Industrial Physics
Plastic cuvette for LICO
Supplier: Hach
SCREW CAP FOR 5 AND 10 ML VIALS 1 * 228 items
Supplier: Mettler - Toledo
VWR®, Eluent buffers for amino acid analyzer
Supplier: VWR Chemicals
Flow Cells for MinION™ and GridION™ Devices
Supplier: Oxford Nanopore Technologies
Dewar vessels, Agil series
Supplier: Air Liquide
Accessories for titrators
Supplier: Mettler - Toledo
Density meters, Densito and DensitoPro
Supplier: Mettler - Toledo