Orders LinkContactUsLinkComponent
Slide boxes
Slide boxes
Slide box, Black, Wood/cardboard, 100, 270×190×35 mm
Catalog # 630-2775
Supplier:   MARIENFELD
Slide boxes
Catalog # 630-2775
Supplier:  MARIENFELD
Slide boxes
Slide boxes
Slide box, Black, Wood/cardboard, 100, 270×190×35 mm
Price
27,30 €
Log in for availability
1 * 1 ST
Product Details & Documents
Slide boxes with numbered slots and index card.
Specifications
  • No. of slides:
    100
  • Colour:
    Black
  • Description:
    Slide box
  • Material:
    Wood/cardboard
  • Seq:
    004
  • W×D×H:
    270×190×35 mm