Orders LinkContactUsLinkComponent
Cylinders, without lid
Cylinders, without lid
180 mm, 180 mm
Catalog # 216-4292
Cylinders, without lid
Catalog # 216-4292
Cylinders, without lid
Cylinders, without lid
180 mm, 180 mm
Price
34,10 €
Log in for availability
1 * 1 ST
Product Details & Documents
Jars, made of glass or stainless steel, with base, without lids.
Suitable for storing dressing material, cotton, clinical thermometers, forceps and other instruments.
Ordering information: Lids with knob available must be ordered separately.
Specifications
  • Ext.Ø:
    180 mm
  • Height:
    180 mm
  • Seq:
    08