Order Entry
Belgium
Orders LinkContactUsLinkComponent
Cylinders, without lid

Cylinders, without lid

300 mm, 100 mm
Catalog # 216-4291
Cylinders, without lid
Catalog # 216-4291
Cylinders, without lid
Cylinders, without lid

300 mm, 100 mm

Price
24,50 €
Log in for availability
1 * 1 ST
Product Details & Documents

Jars, made of glass or stainless steel, with base, without lids.

Suitable for storing dressing material, cotton, clinical thermometers, forceps and other instruments.

Ordering information: Lids with knob available must be ordered separately.

Specifications
  • Ext.Ø:
    100 mm
  • Height:
    300 mm
  • Seq:
    11
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.