Orders LinkContactUsLinkComponent
Labels for slides
Labels for slides
Slide labels on sheets, custom imprint, Paper, Black on white, 11×22 mm
Catalog # 630-1838
Labels for slides
Catalog # 630-1838
Labels for slides
Labels for slides
Slide labels on sheets, custom imprint, Paper, Black on white, 11×22 mm
Price
119,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents
These thin labels are suitable for use with microscopy slides.
  • Permanent adhesive
  • No curling or detaching
Some models can be customised: Imprint can be on the top or bottom of the label
Specifications
  • Colour:
    Black on white
  • Description:
    Slide labels on sheets, custom imprint
  • Dimensions:
    11×22 mm
  • Material:
    Paper
  • Seq:
    002