Orders LinkContactUsLinkComponent
Labels for slides
Slide labels on sheets, custom imprint, Paper, Black on white, 22×22 mm
  630-1839
Labels for slides
  630-1839
Labels for slides
Labels for slides
Slide labels on sheets, custom imprint, Paper, Black on white, 22×22 mm
125,00 €
1 * 1.000 ST
These thin labels are suitable for use with microscopy slides.
  • Permanent adhesive
  • No curling or detaching
Some models can be customised: Imprint can be on the top or bottom of the label
  • Colour:
    Black on white
  • Description:
    Slide labels on sheets, custom imprint
  • Dimensions:
    22×22 mm
  • Material:
    Paper
  • Seq:
    004