Orders LinkContactUsLinkComponent
Test kits, digital titration method
Test kits, digital titration method
Carbon dioxide reagent kit, 10 - 1000 mg/l, 100 Tests
Catalog # HACH2272700
Supplier:   Hach
Test kits, digital titration method
Catalog # HACH2272700
Supplier:  Hach
CAS Number:  
Test kits, digital titration method
Test kits, digital titration method
Carbon dioxide reagent kit, 10 - 1000 mg/l, 100 Tests
CAS Number:  
Price
107,00 €
Log in for availability
1 * 1 EA
Product Details & Documents
Digital titration provides a more precise and accurate method than the drop test method.
  • Reagents are pre-prepared
  • Exact comparison of colour
  • Rugged carrying case for analysis in the field
Caution: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Specifications
Specifications
  • Pk:
    100 Tests
  • Description:
    Carbon dioxide reagent kit
  • Range:
    10 - 1000 mg/l
Specifications