Order Entry
Belgium
Orders LinkContactUsLinkComponent
Tissue processing cassettes, Histosette I

Tissue processing cassettes, Histosette I

Yellow
Catalog # DELT454143
Supplier:   DELTALAB
Tissue processing cassettes, Histosette I
Catalog # DELT454143
Supplier:  DELTALAB
Tissue processing cassettes, Histosette I

Yellow

Price
118,00 €
Log in for availability
1 * 500 ST
Product Details & Documents

Made of acetal polymer. With one-piece integral lid. To lock the cassette, just snap apart the lid, turn it round and lock it into the base of the cassette.

  • Writing area at 30° angle

Documents

Specifications
  • Colour:
    Yellow
  • Seq:
    03