Order Entry
Belgium
Orders LinkContactUsLinkComponent
Plain swabs

Plain swabs

Plain swabs, Wood/cotton, 2,2 mm, 80 mm
N° de catalogue 115-0504
Fournisseur:   BAILLY
Plain swabs
N° de catalogue 115-0504
Fournisseur:  BAILLY
Plain swabs
Plain swabs

Plain swabs, Wood/cotton, 2,2 mm, 80 mm

Prix
10,00 €
Log in for availability
1 * 100 ST
Product Details & Documents

Plain swabs suitable for a wide range of applications.

Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904

Specifications
  • Ø:
    2,2 mm
  • Description:
    Plain swabs
  • Length:
    80 mm
  • Seq:
    001
  • Shaft/head:
    Wood/cotton
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.