Orders LinkContactUsLinkComponent
Hypodermic needles
Hypodermic needles
15, 88,9 mm
Catalog # POPR4090
Supplier:   CADENCE SCIENCE
Hypodermic needles
Catalog # POPR4090
Supplier:  CADENCE SCIENCE
Hypodermic needles
Hypodermic needles
15, 88,9 mm
Price
819,00 €
Log in for availability
1 * 12 Pce
Product Details & Documents
Hypodermic needles are precision made and the points are machine ground to achieve uniform sharpness. The needle cannula are fabricated of type 304 stainless steel tubing. Precision grinding, micro-bead jet blasting, and ultrasonic cleaning assure they are burrs and debris free.
Specifications
  • Gauge:
    15
  • Length:
    88,9 mm
  • Seq:
    02