Orders LinkContactUsLinkComponent
Hypodermic needles
Hypodermic needles
22, 25,4 mm
Catalog # POPR4043
Supplier:   CADENCE SCIENCE
Hypodermic needles
Catalog # POPR4043
Supplier:  CADENCE SCIENCE
Hypodermic needles
Hypodermic needles
22, 25,4 mm
Price
1 170,00 €
Log in for availability
1 * 12 ST
Product Details & Documents
Hypodermic needles are precision made and the points are machine ground to achieve uniform sharpness. The needle cannula are fabricated of type 304 stainless steel tubing. Precision grinding, micro-bead jet blasting, and ultrasonic cleaning assure they are burrs and debris free.
Specifications
  • Gauge:
    22
  • Length:
    25,4 mm