Hypodermic needles
24, 19,1 mm
Catalog # POPR4028
Supplier: CADENCE SCIENCE
Jump to:
Product Details & Documents
Hypodermic needles are precision made and the points are machine ground to achieve uniform sharpness. The needle cannula are fabricated of type 304 stainless steel tubing. Precision grinding, micro-bead jet blasting, and ultrasonic cleaning assure they are burrs and debris free.
Specifications
- Gauge:24
- Length:19,1 mm