Agar, powder, TECHNICAL
Plastic bucket, 5 kg
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Not suitable for microbiological purposes
Synonyms: Agar Agar
- CAS Number:9002-18-0
- MDL Number:MFCD00081288
- Merck Index:13,00184
- Melting Pt:90 °C
- Storage Temperature:Ambient
- EINECS:232-658-1
Certificates
Search by Lot Number10L Plastic bucket for solids UN Y (ZPACK13247010) 2,5L Plastic bottle for solids DIN 80 (ZPACK13252500) 750 ml Plastic bottle for solids DIN 60 (ZPACK13250750)
Specifications
Specification Test Results
- Identification:Passes test
Specifications
- Pack type:Plastic bucket
- Pk:5 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



