Test kits, digital titration method
Total chlorine CN-DT: thiosulphate method, 20 - 2000 mg/l, 1 KIT
Catalog # HACH2471100
Supplier: Hach
Jump to:
Product Details & Documents
Digital titration provides a more precise and accurate method than the drop test method.
- Reagents are pre-prepared
- Exact comparison of colour
- Rugged carrying case for analysis in the field
Caution: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Specifications
Specifications
- Pk:1 KIT
- Description:Total chlorine CN-DT: thiosulphate method
- Range:20 - 2000 mg/l
Specifications
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.