Hydrochloric acid 30%, TECHNICAL
Plastic drum, 20 L
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Synonyms: Hydrochloric acid solution, Muriatic acid
- CAS Number:7647-01-0
- Formula:HCl
- MDL Number:MFCD00011324
- Molecular Weight:36.46 g/mol
- Boiling Pt:110 °C (1013 hPa)
- Melting Pt:–30 °C
- Density:1.18 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-595-7
- UN:1789
- ADR:8,II
- REACH:01-2119484862-27
Documents
Certificates
Search by Lot Number25L Plastic drum for liquids DIN 61 (ZPACK13952500) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951035)
Informatie over de verpakking
P20_VWRC20251.365_SPEC_EN.pdf P20_VWRC20251.445_SPEC_EN.pdf spec_VWRC20251.365 spec_VWRC20251.445
Product specificatie
VWR Chemicals Big Packs and Bulks
Product documentatie
Specifications
Specification Test Results
- Assay:29 - 31 %
Specifications
- Pack type:Plastic drum
- Pk:20 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



