Orders LinkContactUsLinkComponent
Test kits, drop count tests
Test kits, drop count tests
Sulphite reagent set for SU-5 kit, includes sulphamic acid powder pillows, sulphite 1 reagent powder pillows and sulphite 3 solution, up to 200 mg/l SO₃, 1 SET
Catalog # HACH2440000
Supplier:   Hach
Test kits, drop count tests
Catalog # HACH2440000
Supplier:  Hach
CAS Number:  
Test kits, drop count tests
Test kits, drop count tests
Sulphite reagent set for SU-5 kit, includes sulphamic acid powder pillows, sulphite 1 reagent powder pillows and sulphite 3 solution, up to 200 mg/l SO₃, 1 SET
CAS Number:  
Price
137,00 €
Log in for availability
1 * 1 SET
Product Details & Documents
The drop count test is a simple, easy and cost-effective way to test for many parameters like chlorine, chloride, dissolved oxygen, carbon dioxide, sulphite, nitrogen, formaldehyde, and more.
  • Portable - ideal for use in field or in lab
  • Used as a quick reference or for confirmatory test with colour indicator
  • Reagents supplied with the kit
Caution: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Specifications
Specifications
  • Pk:
    1 SET
  • Description:
    Sulphite reagent set for SU-5 kit, includes sulphamic acid powder pillows, sulphite 1 reagent powder pillows and sulphite 3 solution
  • Range:
    up to 200 mg/l SO₃
Specifications
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.