Test kits, drop count tests
Sodium thiosulfate standard solution, stabilized, 0,00246 N, 100 ml marked dropping bottle, 100 mL
Catalog # HACH2408532
Supplier: Hach
Jump to:
Product Details & Documents
The drop count test is a simple, easy and cost-effective way to test for many parameters like chlorine, chloride, dissolved oxygen, carbon dioxide, sulphite, nitrogen, formaldehyde, and more.
- Portable - ideal for use in field or in lab
- Used as a quick reference or for confirmatory test with colour indicator
- Reagents supplied with the kit
: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Specifications
Specifications
- Pk:100 mL
- Description:Sodium thiosulfate standard solution, stabilized, 0,00246 N, 100 ml marked dropping bottle



