Order Entry
Belgium
Orders LinkContactUsLinkComponent
Nitrogen-nitrate standard solution

Nitrogen-nitrate standard solution

Nitrogen-nitrate standard solution, 100 mg/l as NO₃-N (NIST), 500 mL
Catalog # 705-2025
Supplier:   Hach
Nitrogen-nitrate standard solution
Catalog # 705-2025
Supplier:  Hach
CAS Number:  
Nitrogen-nitrate standard solution
Nitrogen-nitrate standard solution

Nitrogen-nitrate standard solution, 100 mg/l as NO₃-N (NIST), 500 mL

CAS Number:  
Price
53,60 €
Log in for availability
1 * 500 mL
Product Details & Documents

Standard solutions for accuracy checks in nitrate nitrogen determinations and also for meter calibrations/standardisation.

Documents

Specifications

Specifications

  • Concentration:
    100 mg/l as NO₃-N (NIST)
  • Pk:
    500 mL
  • Description:
    Nitrogen-nitrate standard solution

Specifications