Orders LinkContactUsLinkComponent
Conductivity standards
Conductivity standards
Sodium chloride standard solution, 995 mg/l TDS, 1000 mg/l Nacl, 1990 µS/cm, 100 mL
Catalog # HACH210542
Supplier:   Hach
Conductivity standards
Catalog # HACH210542
Supplier:  Hach
CAS Number:  
Conductivity standards
Conductivity standards
Sodium chloride standard solution, 995 mg/l TDS, 1000 mg/l Nacl, 1990 µS/cm, 100 mL
CAS Number:  
Price
33,60 €
Log in for availability
1 * 100 mL
Product Details & Documents
The sodium chloride standard solutions are used for calibrating conductivity meters and also used for accuracy checks.
Specifications
Specifications
  • Pk:
    100 mL
  • Description:
    Sodium chloride standard solution, 995 mg/l TDS, 1000 mg/l Nacl
  • Conductivity:
    1990 µS/cm