Large Rectangular Waste Bins
170 L, 720×670×485 mm
CORAG145521
LDPE.
- Mechanical and chemical resistance
- Durability and versatility
- Easy to clean
- Capacity:170 L
- Seq:03
- W×D×H:720×670×485 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



