Test kits, digital titration method
Silver nitrate digital titrator cartridge, 1,128 N, 1
Catalog # HACH1439701
Supplier: Hach
Jump to:
Product Details & Documents
Digital titration provides a more precise and accurate method than the drop test method.
- Reagents are pre-prepared
- Exact comparison of colour
- Rugged carrying case for analysis in the field
Let op: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Specifications
Specifications
- Pk:1
- Description:Silver nitrate digital titrator cartridge, 1,128 N