Orders LinkContactUsLinkComponent
Measuring spoons
Measuring spoon, molded plastic cup, black, 0,2 g
Catalog # HACH63800
Supplier:   Hach
Measuring spoons
Catalog # HACH63800
Supplier:  Hach
Measuring spoons
Measuring spoons
Measuring spoon, molded plastic cup, black, 0,2 g
Price
5,40 €
Log in for availability
1 * 1 ST
Product Details & Documents
Molded plastic cup. Weight based on sodium chloride.
Specifications
  • Capacity:
    0,2 g
  • Description:
    Measuring spoon, molded plastic cup, black
  • Seq:
    03