Order Entry
Belgium
Orders LinkContactUsLinkComponent
Iron standard solution

Iron standard solution

Iron standard solution, pack of 16×10 ml voluette ampoules (NIST), 25 mg/l as Fe, 16
Catalog # HACH1425310
Supplier:   Hach
Iron standard solution
Catalog # HACH1425310
Supplier:  Hach
CAS Number:  
Iron standard solution
Iron standard solution

Iron standard solution, pack of 16×10 ml voluette ampoules (NIST), 25 mg/l as Fe, 16

CAS Number:  
Price
70,10 €
Log in for availability
1 * 16 ST
Product Details & Documents

For accuracy checks in iron determinations.

  • Traceable to NIST
Specifications

Specifications

  • Concentration:
    25 mg/l as Fe
  • Pk:
    16
  • Description:
    Iron standard solution, pack of 16×10 ml voluette ampoules (NIST)

Specifications