Order Entry
Belgium
Orders LinkContactUsLinkComponent
Nitrogen-nitrate standard solution

Nitrogen-nitrate standard solution

Nitrogen-nitrate standard solution, pack of 16×10 ml voluette ampoules, 500 mg/l as NO₃-N (NIST), 16
Catalog # 705-2010
Supplier:   Hach
Nitrogen-nitrate standard solution
Catalog # 705-2010
Supplier:  Hach
CAS Number:  
Nitrogen-nitrate standard solution
Nitrogen-nitrate standard solution

Nitrogen-nitrate standard solution, pack of 16×10 ml voluette ampoules, 500 mg/l as NO₃-N (NIST), 16

CAS Number:  
Price
70,10 €
Log in for availability
1 * 16 ST
Product Details & Documents

Standard solutions for accuracy checks in nitrate nitrogen determinations and also for meter calibrations/standardisation.

Documents

Specifications

Specifications

  • Concentration:
    500 mg/l as NO₃-N (NIST)
  • Pk:
    16
  • Description:
    Nitrogen-nitrate standard solution, pack of 16×10 ml voluette ampoules

Specifications