Orders LinkContactUsLinkComponent
Conductivity standards
Conductivity standards
Conductivity standard solution, 85,47 mg/l TDS, 180 µS/cm, 100 mL
Catalog # HACH2307542
Supplier:   Hach
Conductivity standards
Catalog # HACH2307542
Supplier:  Hach
CAS Number:  
Conductivity standards
Conductivity standards
Conductivity standard solution, 85,47 mg/l TDS, 180 µS/cm, 100 mL
CAS Number:  
Price
32,90 €
Log in for availability
1 * 100 mL
Product Details & Documents
The sodium chloride standard solutions are used for calibrating conductivity meters and also used for accuracy checks.
Specifications
Specifications
  • Pk:
    100 mL
  • Description:
    Conductivity standard solution, 85,47 mg/l TDS
  • Conductivity:
    180 µS/cm
Specifications