Order Entry
Belgium
Orders LinkContactUsLinkComponent
Colour standards

Colour standards

Standard colour solution, 15 Pt-Co units, Bottle, 1× 1 litre, 1 L
Catalog # 707-0009
Supplier:   Hach
Colour standards
Catalog # 707-0009
Supplier:  Hach
CAS Number:  
Colour standards
Colour standards

Standard colour solution, 15 Pt-Co units, Bottle, 1× 1 litre, 1 L

CAS Number:  
Price
177,00 €
Log in for availability
1 * 1 L
Product Details & Documents

For accuracy checks in colour determinations.

  • Simple Accuracy Checks
  • Minimize waste with Voluette ampoules
  • Reliable results

The voluette format has the following advantages: Open a fresh standard amopule for each accuracy check and each ampule contains enough standard for one set of standard additions. Sealed ampoules prevent accidental contamination or evaporation of standard solutions.

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 L
  • Description:
    Standard colour solution, 15 Pt-Co units
  • Pk Info:
    1× 1 litre

Specifications