Order Entry
Belgium
Orders LinkContactUsLinkComponent
Colour standards

Colour standards

Standard colour solution, 500 Pt-Co units, Voluette ampoule, 16× 10 ml, 160 mL
Catalog # 707-0001
Supplier:   Hach
Colour standards
Catalog # 707-0001
Supplier:  Hach
CAS Number:  
Colour standards
Colour standards

Standard colour solution, 500 Pt-Co units, Voluette ampoule, 16× 10 ml, 160 mL

CAS Number:  
Price
91,30 €
Log in for availability
1 * 160 mL
Product Details & Documents

For accuracy checks in colour determinations.

  • Simple Accuracy Checks
  • Minimize waste with Voluette ampoules
  • Reliable results

The voluette format has the following advantages: Open a fresh standard amopule for each accuracy check and each ampule contains enough standard for one set of standard additions. Sealed ampoules prevent accidental contamination or evaporation of standard solutions.

Specifications

Specifications

  • Pack type:
    Voluette ampoule
  • Pk:
    160 mL
  • Description:
    Standard colour solution, 500 Pt-Co units
  • Pk Info:
    16× 10 ml

Specifications