Order Entry
Belgium
Orders LinkContactUsLinkComponent
Phosphorus standard solution

Phosphorus standard solution

Phosphorus standard solution, pack of 16×10 ml voluette ampoules (NIST), 25 mg/ml as P, 16
Catalog # 705-2026
Supplier:   Hach
Phosphorus standard solution
Catalog # 705-2026
Supplier:  Hach
CAS Number:  
Phosphorus standard solution
Phosphorus standard solution

Phosphorus standard solution, pack of 16×10 ml voluette ampoules (NIST), 25 mg/ml as P, 16

CAS Number:  
Price
93,30 €
Log in for availability
1 * 16 ST
Product Details & Documents

Standard solution for accuracy checks in phosphorus determinations.

Specifications

Specifications

  • Concentration:
    25 mg/ml as P
  • Pk:
    16
  • Description:
    Phosphorus standard solution, pack of 16×10 ml voluette ampoules (NIST)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.