Test kits, digital titration method
Digital titrator cartridge assembly, empty, 1
Catalog # HACH1449501
Supplier: Hach
Aller à :
Product Details & Documents
Digital titration provides a more precise and accurate method than the drop test method.
- Reagents are pre-prepared
- Exact comparison of colour
- Rugged carrying case for analysis in the field
Caution: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Spécifications
Spécifications
- Pk:1
- Description:Digital titrator cartridge assembly, empty