Orders LinkContactUsLinkComponent
Test kits, digital titration method
Test kits, digital titration method
Chloride reagent kit: silver nitrate method, 10 - 10000 mg/l, 1 KIT
Catalog # HACH2288000
Supplier:   Hach
Test kits, digital titration method
Catalog # HACH2288000
Supplier:  Hach
CAS Number:  
Test kits, digital titration method
Test kits, digital titration method
Chloride reagent kit: silver nitrate method, 10 - 10000 mg/l, 1 KIT
CAS Number:  
Price
204,00 €
Connexion Pour la disponibilité
1 * 1 KIT
Product Details & Documents
Digital titration provides a more precise and accurate method than the drop test method.
  • Reagents are pre-prepared
  • Exact comparison of colour
  • Rugged carrying case for analysis in the field
Caution: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Spécifications
Spécifications
  • Pk:
    1 KIT
  • Description:
    Chloride reagent kit: silver nitrate method
  • Range:
    10 - 10000 mg/l
Spécifications